Learn More
Biomatik Corporation Mouse Tryptase (TPS) ELISA Kit, 96 tests
Catalog No. 501501615
Encompass_Preferred
Mouse Tryptase (TPS) ELISA Kit for detecting TPS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q02844. Gene ID: 100503895. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1615 Supplier Biomatik Corporation Supplier No. EKL55041
Mouse Tryptase (TPS) ELISA Kit for detecting TPS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q02844. Gene ID: 100503895. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.