Learn More
Biomatik Corporation Human Thymic Stromal Lymphopoietin (TSLP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55049
Encompass_Preferred
Human Thymic Stromal Lymphopoietin (TSLP) ELISA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q969D9. Gene ID: 85480. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1619