Learn More
Biomatik Corporation Mouse Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55053
Encompass_Preferred
Mouse Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit for detecting UBE3A in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.107ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Target Synonyms: ANCR; AS; E6-AP; EPVE6AP; HPVE6A; Papilloma Virus E6-Associated; Angelman Syndrome; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1621