Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor B (VEGFB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55071
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor B (VEGFB) ELISA Kit for detecting VEGFB in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P49766. Gene ID: 22340. Target Synonyms: VEGF-B; VEGFL; VRF; VEGF-Related Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1628