Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor C (VEGFC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55074
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor C (VEGFC) ELISA Kit for detecting VEGFC in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P97953. Gene ID: 22341. Target Synonyms: VEGF-C; Flt4-L; VRPL; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1631