Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55079
Encompass_Preferred
Human Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, saliva, cell lysates, follicular fluid, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P17948. Gene ID: 2321. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1634