Learn More
Biomatik Corporation Human Visfatin (VF) ELISA Kit, 96 tests
Catalog No. 501501640
Encompass_Preferred
Human Visfatin (VF) ELISA Kit for detecting VF in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P43490. Gene ID: 10135. Target Synonyms: Nampt; NAmPRTase; PBEF; PBEF1; Pre-B-Cell Colony Enhancing Factor; Nicotinamide Phosphoribosyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1640 Supplier Biomatik Corporation Supplier No. EKL55089
Human Visfatin (VF) ELISA Kit for detecting VF in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P43490. Gene ID: 10135. Target Synonyms: Nampt; NAmPRTase; PBEF; PBEF1; Pre-B-Cell Colony Enhancing Factor; Nicotinamide Phosphoribosyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.