Learn More
Biomatik Corporation Rat VGF Nerve Growth Factor Inducible (VGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55092
Encompass_Preferred
Rat VGF Nerve Growth Factor Inducible (VGF) ELISA Kit for detecting VGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P20156. Gene ID: 29461. Target Synonyms: NERP1; NERP2; Neuro-Endocrine Specific Protein VGF; Neuroendocrine Regulatory Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1642