Learn More
Biomatik Corporation Human Vasoactive Intestinal Peptide (VIP) ELISA Kit, 96 tests
Catalog No. 501501643
Encompass_Preferred
Human Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.812-500pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 4h, 30min. Accession Number: P01282. Gene ID: 7432. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1643 Supplier Biomatik Corporation Supplier No. EKL55094
Human Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.812-500pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 4h, 30min. Accession Number: P01282. Gene ID: 7432. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.