Learn More
Biomatik Corporation Rat Von Willebrand Factor (vWF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55099
Encompass_Preferred
Rat Von Willebrand Factor (vWF) ELISA Kit for detecting vWF in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000ng/ml. Sensitivity: 6.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: Q62935. Gene ID: 116669. Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1644