Learn More
Biomatik Corporation Mouse Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL55101
Encompass_Preferred
Mouse Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit for detecting WNT5A in tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P22725. Gene ID: 22418. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1646