Learn More
Biomatik Corporation Human 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02004
Encompass_Preferred
The Human 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2) ELISA Kit allows for quantitative determination of 11-Beta-Hydroxysteroid Dehydrogenase Type 2 in Saliva, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80365. Cross Activity: No significant cross-reactivity or interference between 11-Beta-Hydroxysteroid Dehydrogenase Type 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1648