Learn More
Biomatik Corporation Human 17-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD17b1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02007
Encompass_Preferred
The Human 17-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD17b1) ELISA Kit allows for quantitative determination of 17-Beta-Hydroxysteroid Dehydrogenase Type 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14061. Cross Activity: No significant cross-reactivity or interference between 17-Beta-Hydroxysteroid Dehydrogenase Type 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1649