Learn More
Biomatik Corporation Mouse 2', 5'-Oligoadenylate Synthetase 1 (OAS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02014
Encompass_Preferred
Mouse 2',5'-Oligoadenylate Synthetase 1 (OAS1) ELISA Kit for detecting OAS1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11928. Gene ID: 246730. Target Synonyms: 2,5OAS1; 25-OAS1; OAS1; IFI-4; OIAS; OIASI; E18/E16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1651