Learn More
Biomatik Corporation Human 2', 5'-Oligoadenylate Synthetase 2 (OAS2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02016
Encompass_Preferred
The Human 2', 5'-Oligoadenylate Synthetase 2 (OAS2) ELISA Kit allows for quantitative determination of 2', 5'-Oligoadenylate Synthetase 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.30 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29728. Cross Activity: No significant cross-reactivity or interference between 2', 5'-Oligoadenylate Synthetase 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1652