Learn More
Biomatik Corporation Human 2', 5'-Oligoadenylate Synthetase 3 (OAS3) ELISA Kit, 96 tests
Catalog No. 501501653
Encompass_Preferred
The Human 2', 5'-Oligoadenylate Synthetase 3 (OAS3) ELISA Kit allows for quantitative determination of 2', 5'-Oligoadenylate Synthetase 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6K5. Cross Activity: No significant cross-reactivity or interference between 2', 5'-Oligoadenylate Synthetase 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1653 Supplier Biomatik Corporation Supplier No. EKU02017
The Human 2', 5'-Oligoadenylate Synthetase 3 (OAS3) ELISA Kit allows for quantitative determination of 2', 5'-Oligoadenylate Synthetase 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6K5. Cross Activity: No significant cross-reactivity or interference between 2', 5'-Oligoadenylate Synthetase 3 and analogues was observed. Restriction: For Research Use Only.