Learn More
Biomatik Corporation Rat 3-Hydroxy-3-Methylglutaryl Coenzyme A Reductase (HMGCR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02022
Encompass_Preferred
Rat 3-Hydroxy-3-Methylglutaryl Coenzyme A Reductase (HMGCR) ELISA Kit for detecting HMGCR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51639. Gene ID: 25675. Target Synonyms: HMG-CoA Reductase; Hydroxymethylglutaryl-CoA Reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1654