Learn More
Biomatik Corporation 5-Hydroxytryptamine (5-HT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02027
Encompass_Preferred
The 5-Hydroxytryptamine (5-HT) ELISA Kit allows for quantitative determination of 5-Hydroxytryptamine in Serum and Plasma. Species Reactivity: All species. Detection Range: 11.11-900 ng/mL. Sensitivity: 3.97 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between 5-Hydroxytryptamine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1655