Learn More
Biomatik Corporation 5-Methyltetrahydrofolate (5-Me-THF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02032
Encompass_Preferred
The 5-Methyltetrahydrofolate (5-Me-THF) ELISA Kit allows for quantitative determination of 5-Methyltetrahydrofolate in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 1.11-90 ng/mL. Sensitivity: 0.43 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between 5-Methyltetrahydrofolate and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1656