Learn More
Biomatik Corporation Human 5'-Nucleotidase, Ecto (NT5E) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02034
Encompass_Preferred
The Human 5'-Nucleotidase, Ecto (NT5E) ELISA Kit allows for quantitative determination of 5'-Nucleotidase, Ecto in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21589. Cross Activity: No significant cross-reactivity or interference between 5'-Nucleotidase, Ecto and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1657