Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 8 (ADAM8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02051
Encompass_Preferred
The Human A Disintegrin And Metalloprotease 8 (ADAM8) ELISA Kit allows for quantitative determination of A Disintegrin And Metalloprotease 8 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.52 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78325. Cross Activity: No significant cross-reactivity or interference between A Disintegrin And Metalloprotease 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1665