Learn More
Biomatik Corporation Mouse A Disintegrin And Metalloprotease 8 (ADAM8) ELISA Kit, 96 tests
Catalog No. 501501666
Encompass_Preferred
Mouse A Disintegrin And Metalloprotease 8 (ADAM8) ELISA Kit for detecting ADAM8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05910. Gene ID: 11501. Target Synonyms: CD156-A; MS2; CD156a; Disintegrin and metalloproteinase domain-containing protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1666 Supplier Biomatik Corporation Supplier No. EKU02052
Mouse A Disintegrin And Metalloprotease 8 (ADAM8) ELISA Kit for detecting ADAM8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05910. Gene ID: 11501. Target Synonyms: CD156-A; MS2; CD156a; Disintegrin and metalloproteinase domain-containing protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.