Learn More
Biomatik Corporation Human A Disintegrin And Metalloproteinase With Thrombospondin 1 (ADAMTS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02055
Encompass_Preferred
The Human A Disintegrin And Metalloproteinase With Thrombospondin 1 (ADAMTS1) ELISA Kit allows for quantitative determination of A Disintegrin And Metalloproteinase With Thrombospondin 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UHI8. Cross Activity: No significant cross-reactivity or interference between A Disintegrin And Metalloproteinase With Thrombospondin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1667