Learn More
Biomatik Corporation Rat A Disintegrin And Metalloproteinase With Thrombospondin 7 (ADAMTS7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02062
Encompass_Preferred
Rat A Disintegrin And Metalloproteinase With Thrombospondin 7 (ADAMTS7) ELISA Kit for detecting ADAMTS7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q1EHB3. Gene ID: 315879. Target Synonyms: COMPase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1671