Learn More
Biomatik Corporation Mouse Acetyl Coenzyme A Acetyltransferase 1 (ACAT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02068
Encompass_Preferred
Mouse Acetyl Coenzyme A Acetyltransferase 1 (ACAT1) ELISA Kit for detecting ACAT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8QZT1. Gene ID: 110446. Target Synonyms: THIL; ACAT; T2; MAT; Acetoacetyl Coenzyme A Thiolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1674