Learn More
Biomatik Corporation Human Acid Phosphatase 3, Prostatic (ACP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02082
Encompass_Preferred
The Human Acid Phosphatase 3, Prostatic (ACP3) ELISA Kit allows for quantitative determination of Acid Phosphatase 3, Prostatic in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15309. Cross Activity: No significant cross-reactivity or interference between Acid Phosphatase 3, Prostatic and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1679