Learn More
Biomatik Corporation Rat Acid Phosphatase 5, Tartrate Resistant (ACP5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02085
Encompass_Preferred
Rat Acid Phosphatase 5, Tartrate Resistant (ACP5) ELISA Kit for detecting ACP5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29288. Gene ID: 25732. Target Synonyms: TRAP; TrATPase; Tartrate Resistant Acid Phosphatase; Type 5 acid phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1682