Learn More
Biomatik Corporation Human Activated Leukocyte Cell Adhesion Molecule (ALCAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02099
Encompass_Preferred
The Human Activated Leukocyte Cell Adhesion Molecule (ALCAM) ELISA Kit allows for quantitative determination of Activated Leukocyte Cell Adhesion Molecule in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.068 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13740. Cross Activity: No significant cross-reactivity or interference between Activated Leukocyte Cell Adhesion Molecule and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1684