Learn More
Biomatik Corporation Human Activated Protein C (APC) ELISA Kit, 96 tests
Catalog No. 501501685
Encompass_Preferred
The Human Activated Protein C (APC) ELISA Kit allows for quantitative determination of Activated Protein C in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25054. Cross Activity: No significant cross-reactivity or interference between Activated Protein C and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1685 Supplier Biomatik Corporation Supplier No. EKU02100
The Human Activated Protein C (APC) ELISA Kit allows for quantitative determination of Activated Protein C in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25054. Cross Activity: No significant cross-reactivity or interference between Activated Protein C and analogues was observed. Restriction: For Research Use Only.