Learn More
Biomatik Corporation Mouse Activated Protein C (APC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02101
Encompass_Preferred
Mouse Activated Protein C (APC) ELISA Kit for detecting APC in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33587. Gene ID: 19123. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1686