Learn More
Biomatik Corporation Human Adenosine A2b Receptor (ADORA2b) ELISA Kit, 96 tests
Catalog No. 501501693
Encompass_Preferred
The Human Adenosine A2b Receptor (ADORA2b) ELISA Kit allows for quantitative determination of Adenosine A2b Receptor in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29275. Cross Activity: No significant cross-reactivity or interference between Adenosine A2b Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1693 Supplier Biomatik Corporation Supplier No. EKU02129
The Human Adenosine A2b Receptor (ADORA2b) ELISA Kit allows for quantitative determination of Adenosine A2b Receptor in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29275. Cross Activity: No significant cross-reactivity or interference between Adenosine A2b Receptor and analogues was observed. Restriction: For Research Use Only.