Learn More
Biomatik Corporation Human Adenylate Cyclase Activating Polypeptide 1, Pituitary (ADCYAP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02145
Encompass_Preferred
The Human Adenylate Cyclase Activating Polypeptide 1, Pituitary (ADCYAP1) ELISA Kit allows for quantitative determination of Adenylate Cyclase Activating Polypeptide 1, Pituitary in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 pg/mL. Sensitivity: 2.55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P18509. Cross Activity: No significant cross-reactivity or interference between Adenylate Cyclase Activating Polypeptide 1, Pituitary and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1696