Learn More
Biomatik Corporation Cattle Adiponectin (ADP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02150
Encompass_Preferred
The Cattle Adiponectin (ADP) ELISA Kit allows for quantitative determination of Adiponectin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q3Y5Z3. Cross Activity: No significant cross-reactivity or interference between Adiponectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1697