Learn More
Biomatik Corporation Rabbit Adiponectin (ADP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02157
Encompass_Preferred
Rabbit Adiponectin (ADPN) ELISA Kit for detecting ADPN in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.144ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2LDA4. Gene ID: 100009027. Target Synonyms: GBP28; ApM1; AdipoQ; Acrp30; ACDC; APM1; C1Q And Collagen Domain Containing; Adipocyte Complement-Related Protein Of 30 KDa; Adipose Most Abundant Gene Transcript 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1704