Learn More
Biomatik Corporation Human Adiponectin Receptor 1 (ADIPOR1) ELISA Kit, 96 tests
Catalog No. 501501705
Encompass_Preferred
The Human Adiponectin Receptor 1 (ADIPOR1) ELISA Kit allows for quantitative determination of Adiponectin Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96A54. Cross Activity: No significant cross-reactivity or interference between Adiponectin Receptor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1705 Supplier Biomatik Corporation Supplier No. EKU02158
The Human Adiponectin Receptor 1 (ADIPOR1) ELISA Kit allows for quantitative determination of Adiponectin Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96A54. Cross Activity: No significant cross-reactivity or interference between Adiponectin Receptor 1 and analogues was observed. Restriction: For Research Use Only.