Learn More
Biomatik Corporation Human Adipose Differentiation Related Protein (ADRP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02163
Encompass_Preferred
The Human Adipose Differentiation Related Protein (ADRP) ELISA Kit allows for quantitative determination of Adipose Differentiation Related Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q99541. Cross Activity: No significant cross-reactivity or interference between Adipose Differentiation Related Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1707