Learn More
Biomatik Corporation Human Adrenergic Receptor Beta Kinase 1 (ADRbK1) ELISA Kit, 96 tests
Catalog No. 501501709
Encompass_Preferred
The Human Adrenergic Receptor Beta Kinase 1 (ADRbK1) ELISA Kit allows for quantitative determination of Adrenergic Receptor Beta Kinase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.133 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25098. Cross Activity: No significant cross-reactivity or interference between Adrenergic Receptor Beta Kinase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1709 Supplier Biomatik Corporation Supplier No. EKU02174
The Human Adrenergic Receptor Beta Kinase 1 (ADRbK1) ELISA Kit allows for quantitative determination of Adrenergic Receptor Beta Kinase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.133 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25098. Cross Activity: No significant cross-reactivity or interference between Adrenergic Receptor Beta Kinase 1 and analogues was observed. Restriction: For Research Use Only.