Learn More
Biomatik Corporation Mouse Adrenocorticotropic Hormone (ACTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02178
Encompass_Preferred
Mouse Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01193. Gene ID: 18976. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1711