Learn More
Biomatik Corporation Rat Adrenocorticotropic Hormone (ACTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02179
Encompass_Preferred
Rat Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.28-800pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01194. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1712