Learn More
Biomatik Corporation Cattle Adrenomedullin (ADM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02181
Encompass_Preferred
The Cattle Adrenomedullin (ADM) ELISA Kit allows for quantitative determination of Adrenomedullin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O62827. Cross Activity: No significant cross-reactivity or interference between Adrenomedullin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1713