Learn More
Biomatik Corporation Rat Adrenomedullin (ADM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02183
Encompass_Preferred
Rat Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43145. Gene ID: 25026. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1714