Learn More
Biomatik Corporation Mouse Adrenomedullin (ADM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02186
Encompass_Preferred
Mouse Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.29pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P97297. Gene ID: 11535. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1716