Learn More
Biomatik Corporation Human Adrenomedullin 2 (ADM2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02190
Encompass_Preferred
The Human Adrenomedullin 2 (ADM2) ELISA Kit allows for quantitative determination of Adrenomedullin 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 246.9-20000 pg/mL. Sensitivity: 89.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q7Z4H4. Cross Activity: No significant cross-reactivity or interference between Adrenomedullin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1717