Learn More
Biomatik Corporation Human Advanced Glycosylation End Product Specific Receptor (AGER) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02194
Encompass_Preferred
The Human Advanced Glycosylation End Product Specific Receptor (AGER) ELISA Kit allows for quantitative determination of Advanced Glycosylation End Product Specific Receptor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q15109. Cross Activity: No significant cross-reactivity or interference between Advanced Glycosylation End Product Specific Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1720