Learn More
Biomatik Corporation Rabbit Aggrecan (AGC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02203
Encompass_Preferred
Rabbit Aggrecan (AGC) ELISA Kit for detecting AGC in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.244ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28670. Target Synonyms: ACAN; AGC1; AGCAN; CSPG1; CSPCP; CSPGCP; MSK16; SEDK; Cartilage-specific proteoglycan core protein; Large Aggregating Proteoglycan; Chondroitin Sulfate Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1725