Learn More
Biomatik Corporation Human Alanine Aminotransferase (ALT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02209
Encompass_Preferred
The Human Alanine Aminotransferase (ALT) ELISA Kit allows for quantitative determination of Alanine Aminotransferase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24298. Cross Activity: No significant cross-reactivity or interference between Alanine Aminotransferase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1729