Learn More
Biomatik Corporation Mouse Alanine Aminotransferase (ALT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02210
Encompass_Preferred
Mouse Alanine Aminotransferase (ALT) ELISA Kit for detecting ALT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8QZR5. Gene ID: 76282. Target Synonyms: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1730