Learn More
Biomatik Corporation Rat Alanine Aminotransferase (ALT) ELISA Kit, 96 tests
Catalog No. 501501731
Encompass_Preferred
Rat Alanine Aminotransferase (ALT) ELISA Kit for detecting ALT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25409. Gene ID: 81670. Target Synonyms: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1731 Supplier Biomatik Corporation Supplier No. EKU02211
Rat Alanine Aminotransferase (ALT) ELISA Kit for detecting ALT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25409. Gene ID: 81670. Target Synonyms: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.