Learn More
Biomatik Corporation Human Albumin (ALB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02214
Encompass_Preferred
The Human Albumin (ALB) ELISA Kit allows for quantitative determination of Albumin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 246.9-20,000 ng/mL. Sensitivity: 89.6 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P02768. Cross Activity: No significant cross-reactivity or interference between Albumin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1732