Learn More
Biomatik Corporation Human Alkaline Phosphatase, Intestinal (ALPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02244
Encompass_Preferred
The Human Alkaline Phosphatase, Intestinal (ALPI) ELISA Kit allows for quantitative determination of Alkaline Phosphatase, Intestinal in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.63 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09923. Cross Activity: No significant cross-reactivity or interference between Alkaline Phosphatase, Intestinal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1738